The Ambion® MagMAX™ family of products is designed to provide consistent and reliable nucleic acid purification using silica–magnetic particle technology. Magnetic beads offer many benefits for isolating RNA from cells, tissue, or blood and viral RNA from cell-free samples. The technology at the core of the MagMAX™ product line:
- Binds RNA more efficiently than silica-membrane methods
- Offers higher, more consistent RNA yields
- Allows RNA to be eluted in as little as 20 µL of nuclease-free water, resulting in more concentrated RNA